Assay Details
| Gene Symbol: Acvr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007395.3 | NP_031421.1 | 2 - 3 | 357 | 75 | |
| GenBank mRNA | AK154578.1 | - | 379 | 75 | ||
| AK157910.1 | - | 2 - 3 | 358 | 75 | ||
| BC059832.1 | - | 350 | 75 | |||
| BC145775.1 | - | 2 - 3 | 410 | 40131078 | 75 | |
| BC145777.1 | - | 2 - 3 | 410 | 40131080 | 75 | |
| Z31663.1 | - | 2 - 3 | 346 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr1b
Genomic Map