Assay Details
| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007400.2 | NP_031426.2 | 8 - 9 | 964 | 83 | |
| GenBank mRNA | D50411.1 | - | 8 - 9 | 964 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adam12
Genomic Map