Assay Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007402.2 | NP_031428.2 | 11 - 12 | 1108 | 81 | |
| GenBank mRNA | AF013107.1 | - | 11 - 12 | 1094 | 81 | |
| AK136692.1 | - | 11 - 12 | 1107 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam7
Genomic Map