Assay Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007402.2 | NP_031428.2 | 21 - 22 | 2278 | 78 | |
| GenBank mRNA | AF013107.1 | - | 21 - 22 | 2261 | 78 | |
| AK136692.1 | - | 21 - 22 | 2277 | 78 | ||
| AK164965.1 | - | 7 - 8 | 798 | 78 | ||
More Information
Set Membership: |
|

Adam7
Genomic Map