Assay Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007405.2 | NP_031431.2 | 9 - 10 | 1960 | 62 | |
| GenBank mRNA | AK147516.1 | - | 9 - 10 | 1960 | 62 | |
| BC145101.1 | - | 8 - 9 | 1793 | 62 | ||
| M93422.1 | - | 8 - 9 | 1892 | 62 | ||
| M96653.1 | - | 8 - 9 | 1782 | 62 | ||
More Information
Set Membership: |
|

Adcy6
Genomic Map