Assay Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007405.2 | NP_031431.2 | 17 - 18 | 2813 | 96 | |
| GenBank mRNA | AK122279.1 | - | 3 - 4 | 757 | 96 | |
| AK147516.1 | - | 17 - 18 | 2813 | 96 | ||
| BC145101.1 | - | 16 - 17 | 2611 | 96 | ||
| M93422.1 | - | 16 - 17 | 2745 | 96 | ||
| M96653.1 | - | 16 - 17 | 96 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy6
Genomic Map