Assay Details
| Gene Symbol: Adssl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007421.2 | NP_031447.1 | 5 - 6 | 539 | 68 | |
| GenBank mRNA | AK089520.1 | - | 5 - 6 | 514 | 68 | |
| AK146907.1 | - | 5 - 6 | 542 | 68 | ||
| AK149634.1 | - | 5 - 6 | 513 | 68 | ||
| AK150874.1 | - | 5 - 6 | 510 | 68 | ||
| BC039943.1 | - | 5 - 6 | 570 | 4014672 | 68 | |
| M74495.1 | - | 5 - 6 | 497 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adssl1
Genomic Map