Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026646.mm.1 |
NM_001077264.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026646.mm.1 |
NM_001077264.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026646.mm.1 |
NM_007458.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026646.mm.1 |
NM_007458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026646.mm.1 |
TC0700002978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026645.mm.1 |
NM_001077264.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026645.mm.1 |
NM_001077264.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026645.mm.1 |
NM_007458.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026645.mm.1 |
NM_007458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026645.mm.1 |
TC0700002978.mm.1 |
MTA TC |