Assay Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077264.1 | NP_001070732.1 | 11 - 12 | 1603 | 72 | |
| NM_007458.2 | NP_031484.1 | 11 - 12 | 1603 | 72 | ||
| GenBank mRNA | AK135424.1 | - | 11 - 12 | 1532 | 72 | |
| AK155515.1 | - | 11 - 12 | 1540 | 72 | ||
| AK198323.1 | - | 1 - 2 | 38 | 72 | ||
| AK201691.1 | - | 38 | 72 | |||
| BC031433.1 | - | 11 - 12 | 1553 | 4481643 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap2a1
Genomic Map