Assay Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007504.2 | NP_031530.2 | 17 - 18 | 2675 | 80 | |
| GenBank mRNA | AK132537.1 | - | 2 - 3 | 392 | 80 | |
| AY081946.1 | - | 17 - 18 | 2706 | 80 | ||
| BC036292.1 | - | 17 - 18 | 2675 | 4191741 | 80 | |
| X67140.1 | - | 1 - 2 | 88 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2a1
Genomic Map