Assay Details
| Gene Symbol: Baat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007519.3 | NP_031545.2 | 1 - 2 | 237 | 76 | |
| GenBank mRNA | AK017923.1 | - | 1 - 2 | 186 | 76 | |
| BC012683.1 | - | 1 - 2 | 184 | 76 | ||
| U95215.1 | - | 1 - 2 | 145 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK017923.1,NM_007519.3,U95215.1 transcripts.
Set Membership: |
|

Baat
Genomic Map