Assay Details
| Gene Symbol: Bach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007520.2 | NP_031546.1 | 3 - 4 | 1748 | 89 | |
| GenBank mRNA | AK030871.1 | - | 3 - 4 | 1698 | 89 | |
| AK140765.1 | - | 3 - 4 | 1749 | 89 | ||
| AK140903.1 | - | 3 - 4 | 1749 | 89 | ||
| AK141405.1 | - | 3 - 4 | 1683 | 89 | ||
| BC057894.1 | - | 3 - 4 | 1669 | 4978638 | 89 | |
| D86603.1 | - | 3 - 4 | 1758 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bach1
Genomic Map