Assay Details
| Gene Symbol: Hsd3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147114.1 | - | 2 - 3 | 313 | 84 | |
| BC052659.1 | - | 1 - 2 | 54 | 30049155 | 84 | |
| M58567.1 | - | 1 - 2 | 67 | 84 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK147114.1,BC052659.1,M58567.1 transcripts.
Set Membership: |
|

Hsd3b1
Genomic Map