Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 7 - 8 | 901 | 90 | |
| GenBank mRNA | AB119195.1 | - | 5 - 6 | 399 | 90 | |
| AB221614.1 | - | 5 - 6 | 406 | 90 | ||
| AK052726.1 | - | 2 - 3 | 209 | 90 | ||
| AY521462.1 | - | 5 - 6 | 399 | 90 | ||
| AY521463.1 | - | 5 - 6 | 399 | 90 | ||
| BC050841.1 | - | 7 - 8 | 901 | 90 | ||
| BC138535.1 | - | 6 - 7 | 478 | 8861555 | 90 | |
| BC138536.1 | - | 6 - 7 | 478 | 8861556 | 90 | |
| BC145317.1 | - | 7 - 8 | 495 | 90 | ||
| M57999.1 | - | 7 - 8 | 690 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK052726.1 transcripts.
Set Membership: |
|

Nfkb1
Genomic Map