Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 13 - 14 | 1709 | 70 | |
| GenBank mRNA | AB119195.1 | - | 11 - 12 | 1207 | 70 | |
| AB221614.1 | - | 11 - 12 | 1214 | 70 | ||
| AK089660.1 | - | 1 - 2 | 83 | 70 | ||
| AK159423.1 | - | 1 - 2 | 140 | 70 | ||
| AY521462.1 | - | 11 - 12 | 1207 | 70 | ||
| AY521463.1 | - | 11 - 12 | 1207 | 70 | ||
| BC138535.1 | - | 12 - 13 | 1286 | 8861555 | 70 | |
| BC138536.1 | - | 12 - 13 | 1286 | 8861556 | 70 | |
| BC145317.1 | - | 13 - 14 | 1303 | 70 | ||
| M57999.1 | - | 13 - 14 | 1498 | 70 | ||
| S66656.1 | - | 1 - 2 | 253 | 70 | ||
| S89033.1 | - | 1 - 2 | 301 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map