Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 14 - 15 | 1794 | 81 | |
| GenBank mRNA | AB119195.1 | - | 12 - 13 | 1292 | 81 | |
| AB221614.1 | - | 12 - 13 | 1299 | 81 | ||
| AK089660.1 | - | 2 - 3 | 168 | 81 | ||
| AK159423.1 | - | 2 - 3 | 225 | 81 | ||
| AY521462.1 | - | 12 - 13 | 1292 | 81 | ||
| AY521463.1 | - | 12 - 13 | 1292 | 81 | ||
| BC138535.1 | - | 13 - 14 | 1371 | 8861555 | 81 | |
| BC138536.1 | - | 13 - 14 | 1371 | 8861556 | 81 | |
| BC145317.1 | - | 14 - 15 | 1388 | 81 | ||
| M57999.1 | - | 14 - 15 | 1583 | 81 | ||
| S66656.1 | - | 2 - 3 | 338 | 81 | ||
| S89033.1 | - | 2 - 3 | 386 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map