Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 22 - 23 | 2911 | 74 | |
| GenBank mRNA | AB119195.1 | - | 20 - 21 | 2409 | 74 | |
| AB221614.1 | - | 20 - 21 | 2416 | 74 | ||
| AK089660.1 | - | 10 - 11 | 1285 | 74 | ||
| AK146389.1 | - | 8 - 9 | 1130 | 74 | ||
| AK157915.1 | - | 7 - 8 | 911 | 74 | ||
| AK159423.1 | - | 10 - 11 | 1342 | 74 | ||
| AY521462.1 | - | 20 - 21 | 2409 | 74 | ||
| AY521463.1 | - | 20 - 21 | 2409 | 74 | ||
| BC138535.1 | - | 21 - 22 | 2488 | 8861555 | 74 | |
| BC138536.1 | - | 21 - 22 | 2488 | 8861556 | 74 | |
| BC145317.1 | - | 22 - 23 | 2505 | 74 | ||
| L28117.1 | - | 10 - 11 | 73 | |||
| M57999.1 | - | 22 - 23 | 2700 | 74 | ||
| S66656.1 | - | 10 - 11 | 1455 | 74 | ||
| S89033.1 | - | 10 - 11 | 1503 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map