Assay Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290368.1 | NP_001277297.1 | 6 - 7 | 660 | 109 | |
| NM_001290369.1 | NP_001277298.1 | 5 - 6 | 642 | 109 | ||
| NM_001290370.1 | NP_001277299.1 | 4 - 5 | 599 | 109 | ||
| NM_008708.2 | NP_032734.1 | 6 - 7 | 692 | 109 | ||
| GenBank mRNA | AF043327.1 | - | 6 - 7 | 616 | 109 | |
| AK134521.1 | - | 6 - 7 | 656 | 109 | ||
| AK161490.1 | - | 6 - 7 | 660 | 109 | ||
| AK168171.1 | - | 6 - 7 | 660 | 109 | ||
| AK168190.1 | - | 6 - 7 | 660 | 109 | ||
| BC037647.1 | - | 5 - 6 | 589 | 5253732 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmt2
Genomic Map