Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 4 - 5 | 916 | 57 | |
| GenBank mRNA | AK039397.1 | - | 4 - 5 | 628 | 57 | |
| AK044364.1 | - | 4 - 5 | 859 | 57 | ||
| BC048247.1 | - | 4 - 5 | 895 | 57 | ||
| BC058803.1 | - | 4 - 5 | 863 | 57 | ||
| BC129923.1 | - | 4 - 5 | 846 | 40111882 | 57 | |
| BC129924.1 | - | 4 - 5 | 845 | 57 | ||
| X55968.1 | - | 4 - 5 | 849 | 57 | ||
| X60133.1 | - | 4 - 5 | 856 | 57 | ||
| X87952.1 | - | 4 - 5 | 856 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde6b
Genomic Map