Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 10 - 11 | 1467 | 88 | |
| GenBank mRNA | AK039397.1 | - | 10 - 11 | 1179 | 88 | |
| AK044364.1 | - | 10 - 11 | 1410 | 88 | ||
| BC048247.1 | - | 10 - 11 | 1446 | 88 | ||
| BC129923.1 | - | 10 - 11 | 1397 | 40111882 | 88 | |
| BC129924.1 | - | 10 - 11 | 1396 | 88 | ||
| X55968.1 | - | 10 - 11 | 1400 | 88 | ||
| X60133.1 | - | 10 - 11 | 1407 | 88 | ||
| X87952.1 | - | 10 - 11 | 1407 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde6b
Genomic Map