Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 20 - 21 | 2414 | 77 | |
| GenBank mRNA | AK039397.1 | - | 20 - 21 | 2126 | 77 | |
| AK044364.1 | - | 20 - 21 | 2357 | 77 | ||
| BC048247.1 | - | 20 - 21 | 2393 | 77 | ||
| BC058803.1 | - | 16 - 17 | 3761 | 77 | ||
| BC129923.1 | - | 20 - 21 | 2344 | 40111882 | 77 | |
| BC129924.1 | - | 20 - 21 | 2343 | 77 | ||
| X55968.1 | - | 20 - 21 | 2347 | 77 | ||
| X60133.1 | - | 20 - 21 | 2354 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde6b
Genomic Map