Assay Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037551.1 | - | 3 - 4 | 682 | 84 | |
| AK084527.1 | - | 2 - 3 | 271 | 84 | ||
| AK169431.1 | - | 2 - 3 | 274 | 84 | ||
| BC008519.1 | - | 2 - 3 | 276 | 3592726 | 84 | |
| BC085502.1 | - | 2 - 3 | 78 | 5720681 | 84 | |
| L06039.1 | - | 2 - 3 | 161 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Citations (1)
Thyroid hormone receptor-beta is associated with coronary angiogenesis during pathological cardiac hypertrophy.
- Authors
- Makino A, Suarez J, Wang H, Belke DD, Scott BT, Dillmann WH
- Journal
- Endocrinology - Page No 2008 (Wed Apr 01 00:00:00 EDT 2009)
- Volume (Year)
- 150 (2009)
- Product Usage
- -

Pecam1
Genomic Map