Assay Details
| Gene Symbol: Pepd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008820.2 | NP_032846.2 | 12 - 13 | 997 | 64 | |
| GenBank mRNA | AK010581.1 | - | 12 - 13 | 978 | 64 | |
| AK145843.1 | - | 12 - 13 | 1028 | 64 | ||
| AK168384.1 | - | 12 - 13 | 987 | 64 | ||
| AK210234.1 | - | 2 - 3 | 174 | 64 | ||
| BC086644.1 | - | 12 - 13 | 971 | 6807359 | 64 | |
| BC131945.1 | - | 12 - 13 | 999 | 40130222 | 64 | |
| BC131973.1 | - | 12 - 13 | 999 | 40130250 | 64 | |
| D82983.1 | - | 12 - 13 | 984 | 64 | ||
| U51014.1 | - | 12 - 13 | 1020 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pepd
Genomic Map