Assay Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146687.2 | NP_001140159.1 | 8 - 9 | 1227 | 93 | |
| NM_001293646.1 | NP_001280575.1 | 8 - 9 | 1227 | 93 | ||
| NM_001293647.1 | NP_001280576.1 | 8 - 9 | 1240 | 93 | ||
| NM_008844.3 | NP_032870.2 | 8 - 9 | 1227 | 93 | ||
| GenBank mRNA | AB006916.1 | - | 8 - 9 | 1210 | 93 | |
| AK014403.1 | - | 8 - 9 | 1193 | 93 | ||
| AK041402.1 | - | 8 - 9 | 1213 | 93 | ||
| AK154816.1 | - | 8 - 9 | 1219 | 93 | ||
| AK155371.1 | - | 8 - 9 | 1203 | 93 | ||
| AK170286.1 | - | 8 - 9 | 1219 | 93 | ||
| AK171576.1 | - | 8 - 9 | 1201 | 93 | ||
| AK182986.1 | - | 1 - 2 | 90 | 93 | ||
| BC019138.1 | - | 8 - 9 | 1176 | 5041431 | 93 | |
| BC094665.1 | - | 8 - 9 | 1199 | 30356181 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1c
Genomic Map