Assay Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293707.1 | NP_001280636.1 | 5 - 6 | 926 | 63 | |
| NM_008847.3 | NP_032873.2 | 5 - 6 | 923 | 63 | ||
| GenBank mRNA | AK098097.1 | - | 5 - 6 | 880 | 63 | |
| AK150898.1 | - | 5 - 6 | 883 | 63 | ||
| AK151984.1 | - | 5 - 6 | 883 | 63 | ||
| AK158062.1 | - | 5 - 6 | 868 | 63 | ||
| BC003763.1 | - | 5 - 6 | 562 | 3591003 | 63 | |
| BC031774.1 | - | 5 - 6 | 851 | 4503697 | 63 | |
| D86177.1 | - | 5 - 6 | 713 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1a
Genomic Map