Assay Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008851.4 | NP_032877.1 | 2 - 3 | 855 | 99 | |
| NR_075078.1 | 3 - 4 | 549 | 99 | |||
| GenBank mRNA | AF006467.1 | - | 3 - 4 | 430 | 99 | |
| AK155377.1 | - | 3 - 4 | 505 | 99 | ||
| BC044893.1 | - | 2 - 3 | 360 | 4481550 | 99 | |
| BC048150.1 | - | 2 - 3 | 509 | 5706382 | 99 | |
| Y08922.1 | - | 2 - 3 | 856 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm1
Genomic Map