Assay Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008851.4 | NP_032877.1 | 5 - 6 | 1530 | 68 | |
| NR_075078.1 | 6 - 7 | 1224 | 68 | |||
| GenBank mRNA | AF006467.1 | - | 6 - 7 | 1105 | 68 | |
| AK155377.1 | - | 6 - 7 | 1180 | 68 | ||
| BC044893.1 | - | 5 - 6 | 1035 | 4481550 | 68 | |
| BC048150.1 | - | 5 - 6 | 1184 | 5706382 | 68 | |
| Y08922.1 | - | 5 - 6 | 1531 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm1
Genomic Map