Assay Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008851.4 | NP_032877.1 | 15 - 16 | 3035 | 93 | |
| NR_075078.1 | 16 - 17 | 2729 | 93 | |||
| GenBank mRNA | AF006467.1 | - | 16 - 17 | 2610 | 93 | |
| AK155377.1 | - | 15 - 16 | 3508 | 93 | ||
| BC044893.1 | - | 15 - 16 | 2540 | 4481550 | 93 | |
| BC048150.1 | - | 15 - 16 | 2689 | 5706382 | 93 | |
| Y08922.1 | - | 15 - 16 | 3036 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm1
Genomic Map