Assay Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008868.3 | NP_032894.2 | 5 - 6 | 640 | 92 | |
| GenBank mRNA | BC029347.1 | - | 4 - 5 | 485 | 4234596 | 92 |
| U18119.1 | - | 4 - 5 | 466 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968099 | U18119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968099 | NM_008868.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352751 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812330 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169894 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123562 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121175 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968099 | U18119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968099 | NM_008868.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427052 | U18119.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427052 | NM_008868.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347503 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169894 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01002373 | - | - |
| Whole Mouse Genome G4122A | A_51_P427052 | U18119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427052 | NM_008868.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105146 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03566940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03566930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03566939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03566920 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03566911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03704511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03256781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03405694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03736949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03405697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03226815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509575 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420584_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509571 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517639 | - | - |
| Mouse Genome 430 2.0 Array | 1420584_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420584_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18781_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18781_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400014144.mm.1 | NM_008868.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014144.mm.1 | NM_008868.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014144.mm.1 | TC0400001732.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014145.mm.1 | NM_008868.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014145.mm.1 | NM_008868.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014145.mm.1 | TC0400001732.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420584_PM_at | - | - |
| Mouse Expression Set 430 | 1420584_at | - | - |
| Murine Genome U74v2 Set | 99326_at | - | - |
| Murine Genome U74v2 Set | 99326_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420584_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420584_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420584_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420584_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 302346 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729585 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2729585 | - | - |
| mouse-6 v1.1 (Illumina) | scl24785.5.1_109-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24785.5.1_109-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2729585 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2729585 | - | - |

Pla2g2c
Genomic Map