Assay Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008872.2 | NP_032898.2 | 2 - 3 | 176 | 63 | |
| GenBank mRNA | AK053476.1 | - | 2 - 3 | 193 | 63 | |
| AK135857.1 | - | 2 - 3 | 167 | 63 | ||
| AK135965.1 | - | 2 - 3 | 168 | 63 | ||
| AK141665.1 | - | 2 - 3 | 167 | 63 | ||
| AK159620.1 | - | 2 - 3 | 167 | 63 | ||
| AK166639.1 | - | 2 - 3 | 167 | 63 | ||
| BC011256.1 | - | 2 - 3 | 173 | 4038404 | 63 | |
| BC057967.1 | - | 2 - 3 | 156 | 4979664 | 63 | |
| BC061508.1 | - | 2 - 3 | 157 | 5352018 | 63 | |
| CJ151826.1 | - | 2 - 3 | 189 | 63 | ||
| J03520.1 | - | 2 - 3 | 159 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plat
Genomic Map