Assay Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008872.2 | NP_032898.2 | 12 - 13 | 1463 | 75 | |
| GenBank mRNA | AK135857.1 | - | 12 - 13 | 1454 | 75 | |
| AK135965.1 | - | 12 - 13 | 1455 | 75 | ||
| AK141665.1 | - | 12 - 13 | 1452 | 75 | ||
| AK159620.1 | - | 12 - 13 | 1455 | 75 | ||
| AK166639.1 | - | 12 - 13 | 75 | |||
| BC011256.1 | - | 12 - 13 | 1460 | 4038404 | 75 | |
| BC057967.1 | - | 12 - 13 | 1443 | 4979664 | 75 | |
| BC061508.1 | - | 12 - 13 | 1444 | 5352018 | 75 | |
| J03520.1 | - | 12 - 13 | 1446 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plat
Genomic Map