Assay Details
| Gene Symbol: Pms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008886.2 | NP_032912.2 | 11 - 12 | 2057 | 77 | |
| GenBank mRNA | AK083049.1 | - | 11 - 12 | 2014 | 77 | |
| AK146365.1 | - | 11 - 12 | 2070 | 77 | ||
| BC058189.1 | - | 11 - 12 | 2065 | 77 | ||
| BC066110.1 | - | 11 - 12 | 2057 | 77 | ||
| BC083502.1 | - | 11 - 12 | 2017 | 77 | ||
| BC086637.1 | - | 11 - 12 | 2033 | 77 | ||
| BC110684.1 | - | 11 - 12 | 2010 | 77 | ||
| BC141287.1 | - | 11 - 12 | 2033 | 9055899 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pms2
Genomic Map