Assay Details
| Gene Symbol: Pms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008886.2 | NP_032912.2 | 12 - 13 | 2224 | 113 | |
| GenBank mRNA | AK146365.1 | - | 12 - 13 | 2237 | 113 | |
| BC066110.1 | - | 12 - 13 | 2224 | 113 | ||
| BC083502.1 | - | 12 - 13 | 2184 | 113 | ||
| BC086637.1 | - | 12 - 13 | 2200 | 113 | ||
| BC139774.1 | - | 12 - 13 | 1658 | 113 | ||
| BC141287.1 | - | 12 - 13 | 2200 | 9055899 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pms2
Genomic Map