Assay Details
| Gene Symbol: Endou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168693.1 | NP_001162164.1 | 8 - 9 | 1109 | 74 | |
| NM_008902.3 | NP_032928.1 | 7 - 8 | 1108 | 74 | ||
| GenBank mRNA | AK132616.1 | - | 8 - 9 | 1050 | 74 | |
| AK169457.1 | - | 6 - 7 | 1199 | 74 | ||
| M95545.1 | - | 7 - 8 | 1108 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Endou
Genomic Map