Assay Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163557.1 | NP_001157029.1 | 4 - 5 | 503 | 73 | |
| NM_008905.2 | NP_032931.2 | 4 - 5 | 739 | 73 | ||
| GenBank mRNA | AK135964.1 | - | 4 - 5 | 774 | 73 | |
| BC051202.1 | - | 4 - 5 | 587 | 73 | ||
| BC086931.1 | - | 4 - 5 | 858 | 73 | ||
| BC100346.1 | - | 4 - 5 | 503 | 30936010 | 73 | |
| U79024.1 | - | 4 - 5 | 524 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppfibp2
Genomic Map