Assay Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163557.1 | NP_001157029.1 | 19 - 20 | 2024 | 99 | |
| NM_008905.2 | NP_032931.2 | 20 - 21 | 2293 | 99 | ||
| GenBank mRNA | AK012152.1 | - | 16 - 17 | 1683 | 99 | |
| AK166921.1 | - | 10 - 11 | 1161 | 99 | ||
| BC003966.1 | - | 5 - 6 | 593 | 99 | ||
| BC055935.1 | - | 9 - 10 | 894 | 99 | ||
| BC100346.1 | - | 19 - 20 | 2024 | 30936010 | 99 | |
| U79024.1 | - | 20 - 21 | 2078 | 99 | ||
More Information
Set Membership: |
|

Ppfibp2
Genomic Map