Assay Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163557.1 | NP_001157029.1 | 22 - 23 | 2363 | 62 | |
| NM_008905.2 | NP_032931.2 | 23 - 24 | 2632 | 62 | ||
| GenBank mRNA | AK012152.1 | - | 19 - 20 | 2021 | 62 | |
| AK166921.1 | - | 13 - 14 | 1500 | 62 | ||
| BC003966.1 | - | 8 - 9 | 929 | 62 | ||
| BC055935.1 | - | 12 - 13 | 1233 | 62 | ||
| BC100346.1 | - | 22 - 23 | 2363 | 30936010 | 62 | |
| U79024.1 | - | 23 - 24 | 2417 | 62 | ||
More Information
Set Membership: |
|

Ppfibp2
Genomic Map