Assay Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015002.1 | - | 7 - 8 | 1070 | 114 | |
| AK133393.1 | - | 7 - 8 | 1091 | 114 | ||
| BC046617.1 | - | 7 - 8 | 896 | 114 | ||
| BC141079.1 | - | 7 - 8 | 911 | 9055691 | 114 | |
| BC145347.1 | - | 6 - 7 | 786 | 114 | ||
| M81475.1 | - | 7 - 8 | 1064 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp3cc
Genomic Map