Assay Details
| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008921.2 | NP_032947.1 | 10 - 11 | 1081 | 124 | |
| GenBank mRNA | AK011456.1 | - | 10 - 11 | 1063 | 124 | |
| AK088557.1 | - | 10 - 11 | 1101 | 124 | ||
| AK089063.1 | - | 10 - 11 | 1106 | 124 | ||
| AK133878.1 | - | 10 - 11 | 1078 | 124 | ||
| AK160831.1 | - | 8 - 9 | 2843 | 124 | ||
| AK169632.1 | - | 10 - 11 | 1078 | 124 | ||
| BC089544.1 | - | 10 - 11 | 1120 | 30504228 | 124 | |
| D13544.1 | - | 10 - 11 | 1081 | 124 | ||
| J04620.1 | - | 10 - 11 | 1140 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim1
Genomic Map