Assay Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008969.4 | NP_032995.1 | 1 - 2 | 139 | 63 | |
| GenBank mRNA | AK046457.1 | - | 1 - 2 | 66 | 63 | |
| AK138432.1 | - | 1 - 2 | 78 | 63 | ||
| AK142741.1 | - | 1 - 2 | 66 | 63 | ||
| AK159907.1 | - | 1 - 2 | 140 | 63 | ||
| AK160886.1 | - | 1 - 2 | 58 | 63 | ||
| AK163609.1 | - | 1 - 2 | 66 | 63 | ||
| AK170664.1 | - | 1 - 2 | 66 | 63 | ||
| BC005573.1 | - | 1 - 2 | 56 | 3968101 | 63 | |
| BC023322.1 | - | 1 - 2 | 48 | 3968136 | 63 | |
| M34141.1 | - | 1 - 2 | 38 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK046457.1,AK138432.1,AK142741.1,AK159907.1,AK163609.1,AK170664.1,BC005573.1,BC023322.1,M34141.1,NM_008969.4 transcripts.
Set Membership: |
|

Ptgs1
Genomic Map