Assay Details
| Gene Symbol: Pxmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008993.2 | NP_033019.2 | 1 - 2 | 282 | 57 | |
| GenBank mRNA | AF309644.1 | - | 1 - 2 | 136 | 57 | |
| AK003118.1 | - | 1 - 2 | 136 | 57 | ||
| BC057975.1 | - | 1 - 2 | 123 | 5055267 | 57 | |
| BY090564.1 | - | 1 - 2 | 282 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978771 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P309184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104049 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177017 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P309184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23978770 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05692859 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104049 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177017 | - | - |
| Whole Mouse Genome G4122A | A_51_P309184 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100952 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104049 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177017 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04239260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04484228 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04239263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23978771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23978765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04484228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04712815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04495728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04913002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04624482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23978746 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532342 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417841_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532339 | - | - |
| Mouse Genome 430 2.0 Array | 1417841_at | - | - |
| Mouse Genome 430 2.0 Array | 1417841_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19301_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19301_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500026416.mm.1 | TC0500002986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026416.mm.1 | NM_008993.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026416.mm.1 | NM_008993.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026417.mm.1 | TC0500002986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026417.mm.1 | NM_008993.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026417.mm.1 | NM_008993.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026420.mm.1 | TC0500002986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026420.mm.1 | NM_008993.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026420.mm.1 | NM_008993.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026421.mm.1 | TC0500002986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500026421.mm.1 | NM_008993.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026421.mm.1 | NM_008993.2 | Refseq |
| Mouse Expression Set 430 | 1417841_PM_at | - | - |
| Mouse Expression Set 430 | 1417841_at | - | - |
| Murine Genome U74v2 Set | 104098_PM_at | - | - |
| Murine Genome U74v2 Set | 104098_at | - | - |
| Murine Genome U74v2 Set | 167381_f_at | - | - |
| Murine Genome U74v2 Set | 167381_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417841_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417841_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417841_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417841_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 926089 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104050563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219748 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219748 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2466947 | - | - |
| mouse-6 v1.1 (Illumina) | scl26229.6.1_58-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0019301.2_74-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650082 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104050563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0019301.2_74-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650082 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104050563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219748 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219748 | - | - |

Pxmp2
Genomic Map