Assay Details
| Gene Symbol: Adam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291890.1 | NP_001278819.1 | 16 - 17 | 1865 | 80 | |
| NM_001291891.1 | NP_001278820.1 | 16 - 17 | 2060 | 80 | ||
| NM_009616.4 | NP_033746.1 | 16 - 17 | 2050 | 80 | ||
| GenBank mRNA | AB096093.1 | - | 5 - 6 | 893 | 80 | |
| AF019887.2 | - | 16 - 17 | 1955 | 80 | ||
| AK147217.1 | - | 16 - 17 | 1973 | 80 | ||
| BC093480.1 | - | 16 - 17 | 2051 | 80 | ||
| D50410.2 | - | 16 - 17 | 1982 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam19
Genomic Map