Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221647.1 | - | 7 - 8 | 862 | 82 | |
| AF090326.1 | - | 946 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aebp2| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221647.1 | - | 7 - 8 | 862 | 82 | |
| AF090326.1 | - | 946 | 82 | |||
Set Membership: |
|