Assay Details
| Gene Symbol: Aire | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271549.1 | NP_001258478.1 | 8 - 9 | 1084 | 81 | |
| NM_001271550.1 | NP_001258479.1 | 8 - 9 | 1075 | 81 | ||
| NM_001271551.1 | NP_001258480.1 | 8 - 9 | 1072 | 81 | ||
| NM_001271552.1 | NP_001258481.1 | 8 - 9 | 1087 | 81 | ||
| NM_001271553.1 | NP_001258482.1 | 8 - 9 | 1084 | 81 | ||
| NM_001271554.1 | NP_001258483.1 | 8 - 9 | 1075 | 81 | ||
| NM_001271555.1 | NP_001258484.1 | 8 - 9 | 1072 | 81 | ||
| NM_001271556.1 | NP_001258485.1 | 8 - 9 | 1087 | 81 | ||
| NM_001271557.1 | NP_001258486.1 | 8 - 9 | 1084 | 81 | ||
| NM_001271558.1 | NP_001258487.1 | 8 - 9 | 1075 | 81 | ||
| NM_001271559.1 | NP_001258488.1 | 8 - 9 | 1072 | 81 | ||
| NM_009646.2 | NP_033776.1 | 8 - 9 | 1087 | 81 | ||
| NR_073358.1 | 9 - 10 | 1142 | 81 | |||
| GenBank mRNA | AF079536.1 | - | 8 - 9 | 1057 | 81 | |
| AF128115.1 | - | 8 - 9 | 1054 | 81 | ||
| AF128116.1 | - | 8 - 9 | 1045 | 81 | ||
| AF128117.1 | - | 8 - 9 | 1042 | 81 | ||
| AF128118.1 | - | 8 - 9 | 1057 | 81 | ||
| AF128119.1 | - | 8 - 9 | 1054 | 81 | ||
| AF128120.1 | - | 8 - 9 | 1045 | 81 | ||
| AF128121.1 | - | 8 - 9 | 1042 | 81 | ||
| AF128122.1 | - | 8 - 9 | 1057 | 81 | ||
| AF128123.1 | - | 8 - 9 | 1054 | 81 | ||
| AF128124.1 | - | 8 - 9 | 1045 | 81 | ||
| AF128125.1 | - | 8 - 9 | 1042 | 81 | ||
| AF128772.1 | - | 8 - 9 | 1000 | 81 | ||
| AF128773.1 | - | 8 - 9 | 1000 | 81 | ||
| AJ132243.1 | - | 8 - 9 | 1000 | 81 | ||
| AJ243821.1 | - | 8 - 9 | 1000 | 81 | ||
| AK131938.1 | - | 1 - 2 | 386 | 81 | ||
| AK153690.1 | - | 9 - 10 | 1138 | 81 | ||
| BC103510.1 | - | 9 - 10 | 1104 | 40039055 | 81 | |
| BC103511.1 | - | 8 - 9 | 1049 | 40039054 | 81 | |
| BC103518.1 | - | 8 - 9 | 1046 | 40039058 | 81 | |
| EU625343.3 | - | 8 - 9 | 1017 | 81 | ||
| JF267399.1 | - | 8 - 9 | 1017 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aire
Genomic Map