Assay Details
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017250.1 | - | 7 - 8 | 647 | 90 | |
BC087927.1 | - | 7 - 8 | 692 | 30504437 | 90 | |
U65021.1 | - | 7 - 8 | 627 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
