Assay Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009673.2 | NP_033803.1 | 2 - 3 | 350 | 86 | |
| GenBank mRNA | AK147740.1 | - | 2 - 3 | 129 | 86 | |
| AK152185.1 | - | 2 - 3 | 185 | 86 | ||
| AK153388.1 | - | 2 - 3 | 131 | 86 | ||
| AK153476.1 | - | 2 - 3 | 114 | 86 | ||
| CX218962.1 | - | 2 - 3 | 306 | 86 | ||
| D63423.1 | - | 2 - 3 | 88 | 86 | ||
| U29396.1 | - | 2 - 3 | 107 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK147740.1,AK152185.1,AK153388.1,AK153476.1,D63423.1,NM_009673.2,U29396.1 transcripts.
Set Membership: |
|

Anxa5
Genomic Map