Assay Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009673.2 | NP_033803.1 | 7 - 8 | 807 | 97 | |
| GenBank mRNA | AK147740.1 | - | 7 - 8 | 586 | 97 | |
| AK152185.1 | - | 7 - 8 | 642 | 97 | ||
| AK153388.1 | - | 7 - 8 | 588 | 97 | ||
| AK153476.1 | - | 7 - 8 | 571 | 97 | ||
| D63423.1 | - | 7 - 8 | 545 | 97 | ||
| U29396.1 | - | 7 - 8 | 564 | 97 | ||
More Information
Set Membership: |
|

Anxa5
Genomic Map