Assay Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009673.2 | NP_033803.1 | 8 - 9 | 871 | 116 | |
| GenBank mRNA | AK147740.1 | - | 8 - 9 | 650 | 116 | |
| AK152185.1 | - | 8 - 9 | 706 | 116 | ||
| AK153388.1 | - | 8 - 9 | 652 | 116 | ||
| AK153476.1 | - | 8 - 9 | 635 | 116 | ||
| BC003716.1 | - | 8 - 9 | 662 | 3488901 | 116 | |
| D63423.1 | - | 8 - 9 | 609 | 116 | ||
| U29396.1 | - | 8 - 9 | 628 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa5
Genomic Map