Assay Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110794.1 | NP_001104264.1 | 2 - 3 | 292 | 84 | |
| NM_009674.3 | NP_033804.2 | 2 - 3 | 407 | 84 | ||
| GenBank mRNA | AK032013.1 | - | 2 - 3 | 183 | 84 | |
| AK041855.1 | - | 2 - 3 | 101 | 84 | ||
| AK077579.1 | - | 2 - 3 | 100 | 84 | ||
| AK134589.1 | - | 2 - 3 | 111 | 84 | ||
| AK149654.1 | - | 2 - 3 | 99 | 84 | ||
| AK151438.1 | - | 2 - 3 | 101 | 84 | ||
| AK161613.1 | - | 2 - 3 | 101 | 84 | ||
| AK167588.1 | - | 2 - 3 | 75 | 84 | ||
| AK169199.1 | - | 2 - 3 | 74 | 84 | ||
| BC008997.1 | - | 2 - 3 | 91 | 3157926 | 84 | |
| CJ163805.1 | - | 2 - 3 | 292 | 84 | ||
| L13129.1 | - | 2 - 3 | 94 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa7
Genomic Map