Assay Details
| Gene Symbol: Akr1b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009731.2 | NP_033861.2 | 6 - 7 | 709 | 120 | |
| GenBank mRNA | AK002705.1 | - | 6 - 7 | 711 | 120 | |
| AK147775.1 | - | 6 - 7 | 738 | 120 | ||
| BC086929.1 | - | 6 - 7 | 682 | 6775941 | 120 | |
| J05663.1 | - | 6 - 7 | 681 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1b7
Genomic Map